ContactUsLinkComponent
Phthalonitrile for synthesis, Sigma-Aldrich®
Toxic
Phthalonitrile for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Toxic
Phthalonitrile for synthesis, Sigma-Aldrich®
Supplier:  Merck
Certificates
Specification Test Results
  • Solubility:
    0.56 g/l (25 °C)
  • Melting Point:
    138 - 141 °C
  • Molar Mass:
    128.13 g/mol
  • Bulk Density:
    500 kg/m3
  • Vapor Pressure:
    0.04 hPa (20 °C)
  • Density:
    1.24 g/cm3 (20 °C)
  • pH:
    7 (H2O)
  • Ignition Point:
    580 °C
Specification Test Results
Related Information
Frequently Bought Together