2,3,4,5-Tetrachlorophenol analytical standard, Supelco®
Catalog # 442281.
Supplier: Merck
CAS Number:
Specifications
- Bal.:100 mg
- Sekvence:2440
- Pk Info:100 mg
Specifications
About this item
:
Boiling Pt:289 °C (1013 hPa)
Melting Pt:116 °C
Density:1,6 g/cm³ (20 °C)
EINECS:225-531-7
UN:2020
ADR:6.1,III
CAS Number:4901-51-3
Formula:C₆H₂Cl₄O