Mercury disposal kit, Mercurisorb™
Catalog # SIAL63879-1EA
Supplier: Merck
Specifications
- Popis:Mercury disposal kit
- Bal.:1
Specifications
About this item
Rapid absorber for safe disposal of mercury.
Delivery: The kit includes disposable gloves, minishovel and brush, disposable pipettes in a compact plastic case.