Racks for digestion tubes
Catalog # GERH12-0269
Supplier: GERHARDT
Specifications
- Poč. otvorů:6
- Type:EG625
- Popis:Insert rack for 250 ml tubes
- Materiál:Nerezová ocel
- Sekvence:02
- Bal.:1
Specifications
About this item
Insert racks made of stainless steel.
- Very good chemical resistance
- Very good temperature resistance
- Easy to clean
This rack is effective for collecting the single digestion tubes like direct distillation.