0
0,00 Kč
Order Entry
Přihlásit se
/
Registrovat
Products
Services
Digital Solutions
Applications
Support
About
Expresní objednávka
Stav objednávky
Váš účet
Košík
Czech Republic
Expresní objednávka
Stav objednávky
Váš účet
Košík
ContactUsLinkComponent
Domů
n-Eicosane
n-Eicosane, Sigma-Aldrich®
n-Eicosane, Sigma-Aldrich®
:
Merck
n-Eicosane, Sigma-Aldrich®
:
Merck
:
112-95-8
:
:
H₃C(CH₂)₁₈CH₃
:
MFCD00009344
:
342,7 °C (1013 hPa)
:
34…37 °C
:
0,932 g/cm³ (20 °C)
:
M-Clarity™ quality level = MQ100
:
Ambient
:
204-018-1
Frequently Bought Together
Aceton ≥99.5%, GPR RECTAPUR®
: VWR Chemicals
Aceton ≥99%, TECHNICAL
: VWR Chemicals
Detergents, liquid, for automated cleaning of glassware, neodisher® FLA
: Dr. Weigert
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
: VWR Chemicals
Acetonitril ≥99.9%, HiPerSolv CHROMANORM®, Isocratic Grade for HPLC
: VWR Chemicals
Acetonitril, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® USP, super gradient grade suitable for UPLC/UHPLC instruments
: VWR Chemicals
2-Propanol ≥98%, TECHNICAL
: VWR Chemicals
Aceton ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
Acetonitril ≥99.9%, LiChrosolv® Reag. Ph. Eur., gradient grade for liquid chromatography, Supelco®
: Merck
Neutralising agent for the automated cleaning of glassware, neodisher® Z
: Dr. Weigert
Xylen (směs izomerů) ≥98.5%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
: VWR Chemicals
n-Hexan ≥99.0%, SPECTRONORM® for spectroscopy
: VWR Chemicals
Etylenglykol ≥99.5% (by GC), EMSURE® Reag. Ph. Eur., Reag. USP p.a.
: Merck
Voda, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
: VWR Chemicals