
Kyselina šťavelová ≥99%, ReagentPlus®, Sigma-Aldrich®
Catalog # SIAL241172-50G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:50 g
- Sekvence:3368
Specifications
About this item
:
Merck Index:13,06980
Boiling Pt:365 °C (1013 hPa)
Melting Pt:99…100 °C
Density:1,9 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:205-634-3
UN:3261
ADR:8,III
:M-Clarity™ quality level = MQ200
:GHS07
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573