Kyselina malonová
Catalog # EHERC14731000
Supplier: EHRENSTORFER
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:0,25 g
- Sekvence:3029
Specifications
About this item
:
Merck Index:13,05732
Boiling Pt:387 °C (1013 hPa)
Melting Pt:136 °C
Density:1,669 g/cm³ (20 °C)
Flash Pt:172 °C
Storage Temperature:Ambient
EINECS:205-503-0
:GHS07
CAS Number:141-82-2
Formula:C₃H₄O₄
MDL Number:MFCD00002707