Cell extract HL60 (non-induced)
Catalog # ENZOBMLSW1010200
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:200 µl
- Popis:Immunoblotting standard, HL60 cell extract (non-induced)
- Sekvence:02
Specifications
About this item
Whole-cell extract of human HL-60 leukemia cells.
- Band with MW=116 kDa, which represents intact PARP, is detected by Western blotting
- Ready to use for SDS-PAGE
- Use approximately 20 µl sample per lane (~75,000 cells)