ContactUsLinkComponent
Propiononitrile for synthesis, Sigma-Aldrich®
 :  Merck
Propiononitrile for synthesis, Sigma-Aldrich®
 :  Merck
  • Solubility:
    99.4 g/l (20 °C)
  • Melting Point:
    -92 °C
  • Molar Mass:
    55.08 g/mol
  • Boiling Point:
    97 °C (1013 hPa)
  • Vapor Pressure:
    50 hPa (20 °C)
  • Flash Point:
    12 °C
  • Refractive Index:
    1.366 (20 °C, 589 nm)
  • Explosion Limit:
    3.1 - 14 %(V)
  • Density:
    0.782 g/cm3 (20 °C)
  • pH:
    10 (50 g/l, H2O, 20 °C)
  • Ignition Point:
    515 °C
  • Hazard Class:
    3
  • Hazard Class Description:
    Flammable Liquid
Frequently Bought Together