L-Arginin hydrochlorid
A1700.1000
:
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
- Environmentally Preferable:
: L(+)-Arginin hydrochlorid
:
:Ambient
:214-275-1
:1119-34-2
:C₆H₁₅ClN₄O₂
:MFCD00064550
:218
:1.42 g/cm³ (20 °C)
:210.66
:13,00785