EDTA dvojsodná sůl dihydrát, Sigma-Aldrich®
Catalog # SIALE4884-100G
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:100 g
- Sekvence:3401
Specifications
About this item
:
Merck Index:13,03543
Melting Pt:255 °C
:M-Clarity™ quality level = MQ200
Storage Temperature:Ambient
EINECS:205-358-3
CAS Number:6381-92-6
Formula:C₁₀H₁₄N₂Na₂O₈·2H₂O
MDL Number:MFCD00150037