Chlorid vápenatý 1 mol/l in aqueous solution, AVS TITRINORM volumetric solution
Catalog # 190464K
Supplier: VWR Chemicals
CAS Number:
Specifications
- Typ balení:Plastová láhev
- Bal.:1 L
- Sekvence:5037
Specifications
Specification Test Results
- Titer (20°C; real value 0.2 % accuracy):0.998 - 1.002 mol/l
- NIST traceable:Confirmed
Specification Test Results
About this item
:
EINECS:233-140-8
:GHS07
Storage Temperature:Ambient
CAS Number:10043-52-4
Formula:CaCl₂
MDL Number:MFCD00010903