Kyselina šťavelová concentrate for dilution to 0.05 mol/l (0.1 N) vodný roztok, DILUT-IT®, Laboratory Reagent, J.T.Baker®
Catalog # 4665.
Supplier: Avantor
CAS Number:
Specifications
- Bal.:210 mL
- Sekvence:2545
Specifications
Specification Test Results
- Titer (N):0.0998-0.1002
Specification Test Results
About this item
:
ADR:8,III
:Dose for preparation of 1 litre of solution.
EINECS:205-634-3
:GHS07
Storage Temperature:Ambient
CAS Number:144-62-7
Formula:C₂H₂O₄
MDL Number:MFCD00002573