Hořčík 1.000 mg/l v zřed. kyselině dusičné (from MgO) ARISTAR® standard pro ICP
Supplier: VWR Chemicals
Certificates
About this item
MgO in HNO₃ 2 to 5%.
Certifications: Traceable to SRM from NIST, manufactured and tested in ISO 17034 / ISO 17025 accredited facilities.
Delivery: Supplied with certificate of analysis.
Related Information
Certificates
Search by Lot Number125 ml Plastic bottle for liquids DIN 24 (ZPACK15150125) 500 ml Plastic bottle for liquids DIN 28 (ZPACK15150500)
Packaging Information
VWR® for Calibrations
Product Literature
Frequently Bought Together