Orders LinkContactUsLinkComponent
Carrying kit for meter and electrodes, Eutech
Carrying kit for meter and electrodes, Eutech
Economy pH carry kit set, incl. plastic carry case, buffer solutions (pH 4.01, pH 7.00), storage solution, deionised (rinse) water, for Eutech pH 5+ / 6+ meters
Katalogové číslo EUTEECECOPHKIT
Dodavatel:   EUTECH
Carrying kit for meter and electrodes, Eutech
Katalogové číslo EUTEECECOPHKIT
Dodavatel:  EUTECH
Carrying kit for meter and electrodes, Eutech
Carrying kit for meter and electrodes, Eutech
Economy pH carry kit set, incl. plastic carry case, buffer solutions (pH 4.01, pH 7.00), storage solution, deionised (rinse) water, for Eutech pH 5+ / 6+ meters
Přejít na:
Price
2 010,00 Kč
Přihlásit se pro dostupnost
1 * 1 SET
Product Details & Documents
Made of sturdy reinforced plastic, the kit comes complete with calibrating sachets and sample bottles, and is thoughtfully designed to hold one meter and electrodes.
Specifikace
  • Popis:
    Economy pH carry kit set, incl. plastic carry case, buffer solutions (pH 4.01, pH 7.00), storage solution, deionised (rinse) water, for Eutech pH 5+ / 6+ meters