UV-Vis wavelenth standard
Catalog # MICSPQA-HOX-01
Supplier: MICROSOLV
CAS Number:
Specifications
- Bal.:30 ml
- Popis:Wavelength calibration standard
Specifications
About this item
Wavelength calibration standard solution for UV-Vis detectors is composed of 4% Holmium oxide in 10% Perchloric acid.