Orders LinkContactUsLinkComponent
Multi-parameter meter, handheld, Professional Plus
Dodavatel:   YSI ENVIRONMENTAL
Multi-parameter meter, handheld, Professional Plus
Dodavatel:  YSI ENVIRONMENTAL
Certifikáty
Multi-parameter meter, handheld, Professional Plus
Multi-parameter meter, handheld, Professional Plus
12 Dostupných variant níže
Product Details & Documents
The Professional Plus is one of the most versatile multi-parameter handheld meters on the market. A combination of up to 11 parameters can be measured on a single bulkhead, including pH, ORP, ammonia, nitrate, chloride, DO, conductivity, salinity, TDS, barometric pressure and temperature, meaning only one cable is required.
  • Rubber, over-moulded IP 67 housing (even without battery cover)
  • Rugged cable sensors with military specification, waterproof connectors
  • USB connection and Data Manager software with GLP functions
  • 5000 data sets memory
  • Easy to read graphic, backlit display and keypad, and multiple language interface: English, German, French, Spanish, Italian, Portuguese and Norwegian
The Quatro cable/bulkhead allows the measuring of up to six parameters at one time (e.g. conductivity, temperature, dissolved oxygen, 2 mV readings and barometric pressure) and has a combined weight of 525 g.
The sensors are totally interchangeable and easily replaceable by the user, and there are a variety of cable lengths available (1, 4, 10, 20 and 30 m). The Pro Plus can be used with other bulkheads to measure one, two or three parameters at the same time.
Three year instrument warranty, two year cable warranty.
 : The instrument Professional Plus is supplied with Data Manager software, USB cable and USB saddle. Quatro cable and sensors need to be ordered separately.
Product Family Options
Quatro cables/bulkhead
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • 665-0203
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 1 m
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 1 m
    01
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 83 330,00 Kč
    83 330,00 Kč
    1 * 1 KS
       83 330,00 Kč
  • 665-0207
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 4 m
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 4 m
    02
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 85 760,00 Kč
    85 760,00 Kč
    1 * 1 KS
       85 760,00 Kč
  • 665-0204
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 10 m
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 10 m
    03
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 88 230,00 Kč
    88 230,00 Kč
    1 * 1 KS
       88 230,00 Kč
  • 665-0205
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 20 m
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 20 m
    04
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 95 540,00 Kč
    95 540,00 Kč
    1 * 1 KS
       95 540,00 Kč
  • 665-0206
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 30 m
    Quatro cable/bulkhead (includes conductivity/temperature sensor), 30 m
    05
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 101 670,00 Kč
    101 670,00 Kč
    1 * 1 KS
       101 670,00 Kč
Sensors
Informace o produktu
  • Sekvence
    Dostupnost
    Price
  • 665-0211
    pH electrode 1001
    pH electrode 1001
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 10 070,00 Kč
    10 070,00 Kč
    1 * 1 KS
       10 070,00 Kč
  • 665-0210
    Polarographic DO sensor
    Polarographic DO sensor
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 10 800,00 Kč
    10 800,00 Kč
    1 * 1 KS
       10 800,00 Kč
  • 665-0209
    Galvanic DO sensor
    Galvanic DO sensor
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 10 800,00 Kč
    10 800,00 Kč
    1 * 1 KS
       10 800,00 Kč
  • 665-0208
    ORP electrode
    ORP electrode
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 12 030,00 Kč
    12 030,00 Kč
    1 * 1 KS
       12 030,00 Kč
  • 665-0212
    Ammonium ion selective electrode (0 - 200 mg/l, 0 to +30 °C)
    Ammonium ion selective electrode (0 - 200 mg/l, 0 to +30 °C)
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 21 590,00 Kč
    21 590,00 Kč
    1 * 1 KS
       21 590,00 Kč
  • 665-0213
    Chloride ion selective electrode (0 - 1000 mg/l, 0 to +40 °C)
    Chloride ion selective electrode (0 - 1000 mg/l, 0 to +40 °C)
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 21 590,00 Kč
    21 590,00 Kč
    1 * 1 KS
       21 590,00 Kč
  • 665-0214
    Nitrate ion selective electrode (0 - 200 mg/l, 0 to +30 °C)
    Nitrate ion selective electrode (0 - 200 mg/l, 0 to +30 °C)
    Přihlásit se pro dostupnost
    Přihlásit se pro dostupnost
    1 * 1 KS
    1 * 1 KS - 21 590,00 Kč
    21 590,00 Kč
    1 * 1 KS
       21 590,00 Kč
Accessories
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Soft-sided carrying case
    Soft-sided carrying case
    664-0025
    15
    Přihlásit se pro dostupnost
    1 * 1 KS - 7 640,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Hard-sided carrying case
    Hard-sided carrying case
    664-0024
    08
    Přihlásit se pro dostupnost
    1 * 1 KS - 17 660,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Bal.
    Dostupnost
    Price
  • Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each)
    Confidence solution 5580, for verifying pH, ORP, and conductivity values, 6 plastic bottles (1 pint each)
    YSIA005580
    100
    1 KIT
    Přihlásit se pro dostupnost
    1 * 1 KIT - 6 130,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Small tripod for flat surface
    Small tripod for flat surface
    665-0069
    14
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 880,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Cable weight, 140 g, attached to stainless steel probe guard
    Cable weight, 140 g, attached to stainless steel probe guard
    664-0026
    04
    Přihlásit se pro dostupnost
    1 * 1 KS - 1 650,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • Flow cell for Quatro cable/bulkhead
    Flow cell for Quatro cable/bulkhead
    665-0235
    07
    Přihlásit se pro dostupnost
    1 * 1 KS - 23 560,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • pH/ORP kit (must be screwed into probe)
    pH/ORP kit (must be screwed into probe)
    665-0091
    11
    Přihlásit se pro dostupnost
    1 * 1 KS - 22 550,00 Kč
Informace o produktu
  • Katalogové číslo
    Sekvence
    Dostupnost
    Price
  • pH kit (must be screwed into probe)
    pH kit (must be screwed into probe)
    665-0090
    10
    Přihlásit se pro dostupnost
    1 * 1 KS - 16 700,00 Kč