parafín, Teplota tuhnutí: 58-62°C, Sigma-Aldrich®
Catalog # SIAL327212-1KG
Supplier: Merck
CAS Number:
Specifications
- Typ balení:Glass bottle for solids
- Bal.:1 kg
- Sekvence:4024
Specifications
About this item
:
:M-Clarity™ quality level = MQ200
EINECS:232-315-6
CAS Number:8002-74-2
:64742-51-4