Ladies coat, LW63
Dodavatel: Portwest
Certifikáty
O tomto produktu
This lab coat made from Kingsmill fabric (65% polyester and 35% cotton) is suitable for a wide range of work environments.
- Concealed stud fastening
- One chest pocket and two side pockets
- Darts for a tailored style
- Practical and comfortable
Weight: 210 g/m²
Specifikace
- Catalog No:
- PRTWLW63WHRXXS
- PRTWLW63WHRXS
- PRTWLW63WHRXXL
- PRTWLW63WHRXL
- PRTWLW63WHRM
- PRTWLW63WHRS
- PRTWLW63WHRL
- Model:
- LW63
- LW63
- LW63
- LW63
- LW63
- LW63
- LW63
- Aplikace oblečení:
- General Purpose, Medical, Laboratory, Clinical, Food
- General Purpose, Medical, Laboratory, Clinical, Food
- General Purpose, Medical, Laboratory, Clinical, Food
- General Purpose, Medical, Laboratory, Clinical, Food
- General Purpose, Medical, Laboratory, Clinical, Food
- General Purpose, Medical, Laboratory, Clinical, Food
- General Purpose, Medical, Laboratory, Clinical, Food
- Rod:
- Pro ženy
- Pro ženy
- Pro ženy
- Pro ženy
- Pro ženy
- Pro ženy
- Pro ženy
- Sleeve or leg length:
- Long
- Long
- Long
- Long
- Long
- Long
- Long
- Typ uzávěru:
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Zapínání na druky
- Tkanina:
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- 65% polyester and 35% cotton
- Hmotnost:
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²
- 210 g/m²