Titan ≥99.99% (metals basis), plate 100x100 mm (3.9x3.9 in), Thickness: 3.175 mm (0.125 in)
Catalog # 43361.KG
Supplier: Thermo Fisher Scientific
CAS Number:
Specifications
- Bal.:1
- Sekvence:0001
Specifications
About this item
:
Merck Index:12,09610
Boiling Pt:3262 °C (1013 hPa)
Melting Pt:1660 °C
Density:4,51 g/cm³ (20 °C)
Storage Temperature:Ambient
EINECS:231-142-3
:GHS02
CAS Number:7440-32-6
Formula:Ti
MDL Number:MFCD00011264