Safety helmets, Vision Plus
Safety helmet, unvented, with ratchet, Bílá
Katalogové číslo CENUS10PLUSEWR
Dodavatel: CENTURION SAFETY PRODUCTS
Přejít na:
Product Details & Documents
The Vision Plus helmet offers wearers of prescription glasses total safety reassurance – with moulded integrated eye protection certified to grade A high impact levels. Helmet shell is made from ABS plastic.
- Optical clarity to Class 1 for continuous use
- Large side pull tabs enable the wearer to quickly and easily move the visor up and down even when wearing gloves
- Moulded design provides improved lower cheek coverage
- Anti-scratch and anti-mist coating for improved durability and visibility
- Wheel ratchet, non-vented
Weight: 525 g with fitted visor
Certifications: Helmet: EN 397; Integrated visor: EN 166
Specifikace
- Barva:Bílá
- Popis:Safety helmet, unvented, with ratchet
Accessories
Informace o produktu
- Katalogové čísloSekvenceDostupnostPrice
- Euro clips also for use on Nexus and Vision Plus helmets, S565Euro clips also for use on Nexus and Vision Plus helmets, S565CENUS56502Přihlásit se pro dostupnost1 * 1 KS - 112,00 Kč