Residual oxygen measuring systems, ResOx 5695-H / ResOx 5695-L
These ready-to-use oxygen measuring systems with gas pump are ideal for the quality control of MAP food packaging and comparable applications.
- With data logger and interface
- Pressure-compensated measurement - particularly important for rigid packages
- Membrane filter prevents unintended suction of particles / liquid
- Independent battery-operated gas pump
- Waterproof device and sensor (IP65, IP67)
ResOx 5695-H: With sensor GOEL 370, for gases with elevated CO2 percentage
ResOx 5695-L: With sensor GOEL 381, for generic protection gases and precise measurements also at low values
Delivery rate: Approx. 80 ml/min (with GDZ 29 Filter)
: The measuring systems are supplied with sensor (GOEL 370 or GOEL 381), gas pump GS 150, battery, connection lines, hoses/T-piece, 2 filters, 2 puncture needles Ø 0,9 mm, 1× 45 pcs. adhesive seal and carrying case GKK 1420.
- Catalog No:
- GRSG610040
- GRSG610041
- Rozpuštěný kyslík - saturace O₂:
- 0,0 - 100,0 %
- 0,0 - 100,0 %
- Rozpuštěný kyslík - rozsah barometrického tlaku:
- 300 - 5000 hPa (Sensor: 500 - 2000 hPa)
- 300 - 5000 hPa (Sensor: 500 - 2000 hPa)
- DO - kalibrace:
- Quick calibration on air at the push of a button or 2-point / 3-point (Air +0 % and 100 %)
- Quick calibration on air at the push of a button or 2-point / 3-point (Air +0 % and 100 %)
- Rozsah teplot:
- 0,0...50,0 °C
- 0,0...50,0 °C
- Displej:
- With backlight
- With backlight
- Kapacita pro ukládání dat:
- Cyclical: 10000, Single: 1000
- Cyclical: 10000, Single: 1000
Frequently Bought Together