Z-Pro-Nle-Asp-CHO
Catalog # ENZOBMLZW94900100
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:100 µG
- Sekvence:02
- Název proteinu/peptidu:Z-Pro-Nle-Asp-CHO
Specifications
About this item
Highly specific aldehyde based inhibitor for the caspase-like peptidic activity of the 20S proteasome.