Z-VAD(OMe)-FMK
ENZOBMLP4160001
:
- Bal.:1 mg
- Sekvence:02
- Název proteinu/peptidu:Z-VAD(OMe)-FMK
- Čistota:>95%
Caspase inhibitor
Irreversible and cell permeable broad-spectrum caspase inhibitor. Inhibits Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP.1 cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine). For cell free assays use Prod. No. ALX-260-020.