ContactUsLinkComponent
Z-VAD(OMe)-FMK
Z-VAD(OMe)-FMK
  ENZOBMLP4160001
 :  
Z-VAD(OMe)-FMK
  ENZOBMLP4160001
 :  
  • Bal.:
    1 mg
  • Sekvence:
    02
  • Název proteinu/peptidu:
    Z-VAD(OMe)-FMK
  • Čistota:
    >95%
Caspase inhibitor
Irreversible and cell permeable broad-spectrum caspase inhibitor. Inhibits Fas-mediated apoptosis in Jurkat T cells and apoptosis in THP.1 cells induced by diverse stimuli (including cycloheximide, thapsigargin, etoposide and staurosporine). For cell free assays use Prod. No. ALX-260-020.