Mu-Phe-hPhe-FMK
Catalog # ENZOBMLEI3230001
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:1 mg
- Sekvence:02
- Název proteinu/peptidu:Mu-Phe-hPhe-FMK
- Čistota:98%
Specifications
About this item
Cathepsin inhibitor
Irreversible, non-toxic, cell-permeable inhibitor of cysteine proteases calpain, cathepsins B and L, and Trypanosome trypanopain. Can be used in vitro (typically 20 µM) or in vivo. Inhibits tumor formation and growth.