Autocamtide-2-related inhibitory peptide
ENZOALX151030MC05
:
- Bal.:0,5 mg
- Sekvence:02
- Název proteinu/peptidu:Autocamtide-2-related inhibitory Peptide
- Čistota:>98%
Cell permeable CaM kinase II inhibitor
Corresponds to Prod. No. ALX-151-029 but this one is N-terminal myristoylated to increase cell permeability.