Proteinase K
Catalog # ENZOENZ33801
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:5 mg
- Sekvence:02
- Formulář:Lyophilised
- Název enzymu:Proteinase K
- Pk Info:2×5 mg
Specifications
About this item
ENZO Proteinase K is specifically designed for use in the pretreatment of formalin-fixed tissue and paraformaldehyde-fixed cells in preparation for in situ hybridization and detection studies.