ContactUsLinkComponent
Proteinase K
Proteinase K
Catalog # ENZOENZ33801
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Proteinase K
Catalog # ENZOENZ33801
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Bal.:
    5 mg
  • Sekvence:
    02
  • Formulář:
    Lyophilised
  • Název enzymu:
    Proteinase K
  • Pk Info:
    2×5 mg
Specifications
About this item
ENZO Proteinase K is specifically designed for use in the pretreatment of formalin-fixed tissue and paraformaldehyde-fixed cells in preparation for in situ hybridization and detection studies.