Mouse recombinant IL-4
Catalog # ENZOENZPRT1330020
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:20 µG
- Typ proteinu/peptidu:Recombinant
- Druhy:Mouse
- Sekvence:02
- Proteinová synonyma:Interleukin-4
- Název proteinu/peptidu:IL-4
- Čistota:>98% (SDS-PAGE gel & HPLC)
- Molekulární hmotnost:~14 kDa
- Formulace:Lyophilized
Specifications
About this item
Produced in E.coli.