Human recombinant p62 (GST-tag),GST Tag
ENZOENZPRT1200050
:
- Bal.:50 µG
- Typ proteinu/peptidu:Recombinant
- Druhy:Human
- Sekvence:02
- Sekvence značek:GST Tag
- Proteinová synonyma:SQSTM1|EBI3-associated protein of 60 kDa|EBIAP|Sequestosome-1
- Název proteinu/peptidu:p62 (GST-tag)
- Čistota:>90% (SDS-PAGE)
- Molekulární hmotnost:~74.4 kDa
- Formulace:Liquid. In 1 M PBS containing 100 mm GSH, 1% Triton X-100, and 15% glycerol
Produced in E. coli. Human p62 (1-440 aa) is fused to a N-terminal GST-tag.