Native N. Gonorrhoeae antigen
ENZOALX6301291000
:
- Bal.:1 mL
- Typ proteinu/peptidu:Native
- Sekvence:02
- Název proteinu/peptidu:N. Gonorrhoeae Antigen
- Čistota:>99%
- Formulace:Liquid, PBS.
- Čištění:Homogenized lysate of pure bacteria.
Bacterial lysate ideal for use as a broad positive control.
From Neisseria gonorrhoeae, strain ATCC 19424. Inactivated by 3 hours at 56°C.