Human recombinant TRAIL (soluble) (from E. coli)
Catalog # ENZOBMLSE7210100
Supplier: Enzo Life Sciences
CAS Number:
Specifications
- Bal.:100 µG
- Typ proteinu/peptidu:Recombinant
- Zdroj:E. coli
- Druhy:Human
- Sekvence:02
- Biologická aktivita:Induces apoptosis in U937 cells at 1 µg/ml, within 5 h, at 0,5 - 2,0 µg/ml, induces apoptosis in various thyroid carcinoma lines within 18 h, user must determine optimal concentrations and time course for induction, please note that this protein does
- Název proteinu/peptidu:TRAIL (soluble)
- Čistota:>95% (SDS-PAGE)
- Formulace:xx mg/ml in 10 mM Na₂HPO₄, 2 mM KH₂PO₄, 2,7 mM KCl, 500 mM NaCl, 10% glycerol, 0,1 mM DTT.
Specifications
About this item
Produced in E. coli. Human TRAIL (aa 114-281).