Human recombinant HDAC2 (full-length), HIS tag
ENZOBMLSE5330050
:
- Bal.:50 µG
- Typ proteinu/peptidu:Recombinant
- Druhy:Human
- Sekvence:02
- Sekvence značek:HIS Tag
- Název proteinu/peptidu:HDAC2 (full-length)
- Čistota:>50% by SDS-PAGE
- Koncentrace:0,68 µg/µl (lot #12021014)
- Formulace:Liquid. In 50 mM TRIS pH 8, containing 138 mM sodium chloride and 10% glycerol.
Highly active
Produced in insect cells.