ContactUsLinkComponent
Human cathepsin G (from Neutrophils)
Catalog # ENZOBMLSE2830100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human cathepsin G (from Neutrophils)
Catalog # ENZOBMLSE2830100
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Bal.:
    100 µG
  • Zdroj:
    Neutrophils
  • Druhy:
    Human
  • Sekvence:
    02
  • Název proteinu/peptidu:
    Cathepsin G
  • Čistota:
    95% by SDS-PAGE
  • Třída:
    Purified
Specifications
About this item
Isolated from human neutrophils. Active enzyme. Negative for HbsAg, anti-HBc, anti-HCV, and anti-HIV-1 and -2.