Human native cathepsin D (from liver)
ENZOBMLSE1990025
:
- Bal.:25 µG
- Typ proteinu/peptidu:Native
- Zdroj:Liver
- Druhy:Human
- Sekvence:02
- Pokyny pro rekonstituce:Reconstitute just before first use by adding 25 - 50 µl of water.
- Název proteinu/peptidu:Cathepsin D
- Čistota:>95% (SDS-PAGE)
- Formulace:Lyophilised with phosphate buffer.
- Třída:Purified
Isolated from human liver.