ContactUsLinkComponent
CDK2 substrate,Biotin
  ENZOBMLP2660001
 :  
CDK2 substrate,Biotin
  ENZOBMLP2660001
 :  
  • Bal.:
    1 mg
  • Spojení:
    Biotin
  • Sekvence:
    02
  • Název proteinu/peptidu:
    CDK2 Substrate
  • Čistota:
    >95% (HPLC)
  • Formulace:
    Lyophilized
This peptide, Biotin-Axh-HHASPRK, is a specific substrate for CDK2. The biotin allows the peptide to be used in kinase assays with streptavidin-bound membranes.