CDK2 substrate,Biotin
ENZOBMLP2660001
:
- Bal.:1 mg
- Spojení:Biotin
- Sekvence:02
- Název proteinu/peptidu:CDK2 Substrate
- Čistota:>95% (HPLC)
- Formulace:Lyophilized
This peptide, Biotin-Axh-HHASPRK, is a specific substrate for CDK2. The biotin allows the peptide to be used in kinase assays with streptavidin-bound membranes.