FGFR substrate
Catalog # ENZOBMLP2440001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:1 mg
- Sekvence:02
- Název proteinu/peptidu:FGFR Substrate
- Čistota:95%
Specifications
About this item
This peptide, KEAPEDLYKDFLTL, is derived from the KDR sequence and is a substrate for FGFR.