ContactUsLinkComponent
Boc-Gln-Ala-Arg-AMC
Boc-Gln-Ala-Arg-AMC
Catalog # ENZOBMLP2370005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Boc-Gln-Ala-Arg-AMC
Catalog # ENZOBMLP2370005
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Specifications
  • Bal.:
    5 mg
  • Sekvence:
    02
  • Název proteinu/peptidu:
    Boc-Gln-Ala-Arg-AMC
  • Čistota:
    >99%
Specifications
About this item
Protease substrate
Fluorogenic substrate for trypsin (kcat/Km=2,0×107 M-1sec-1; Km=6.0 µM); matriptase/MT-SP1 and matriptase-2/TMPRSS6; PRSS12; KLK5; KLK11/PRSS20, KLK12, and KLK14; tryptase and thrombin; and dust mite allergen Der p 1. Ex.: 380 nm, Em.: 460 nm, although the following Ex/Em can also be used: 355,375/440,450. This substrate is useful for inhibitor screening and kinetic analysis. Also available: fluorogenic calibration standard AMC (BML-KI144).