Suc-Ala-Ala-Pro-Phe-pNA
Katalogové číslo ENZOBMLP1410025
Dodavatel: ENZO LIFE SCIENCES
Číslo CAS:
Specifikace
- Bal.:25 mg
- Sekvence:02
- Název proteinu/peptidu:Suc-Ala-Ala-Pro-Phe-pNA
- Čistota:99%
Specifikace
O tomto produktu
Cathepsin substrate
Chromogenic substrate cleaved by cathepsin G, subtilisins, chymotrypsin, chymase, and cyclophilins, but not neutrophil elastase. Release of pNA is monitored at 405 to 410 nm. This substrate is useful for inhibitor screening and kinetic analysis. Also available: calibration standard pNA (BML-KI145).