ContactUsLinkComponent
Mouse recombinant adiponectin
  ENZOALX522059C050
 :  
undefined
Mouse recombinant adiponectin
  ENZOALX522059C050
 :  
  • Bal.:
    50 µG
  • Typ proteinu/peptidu:
    Recombinant
  • Druhy:
    Mouse
  • Sekvence:
    02
  • Biologická aktivita:
    Inhibits serum glucose production in mice. Protects mouse hepatocytes from TNF-α-induced cell death in vitro.
  • Pokyny pro rekonstituce:
    Reconstitute with 100 µl sterile water.
  • Proteinová synonyma:
    ACDC
  • Název proteinu/peptidu:
    Adiponectin
  • Čistota:
    >90% (SDS-PAGE)
  • Koncentrace:
    0,5 mg/ml after reconstitution
  • Formulace:
    Lyophilised. Contains PBS.
Mouse adiponectin (aa 18-247) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.
  • Produced in HEK 293 cells
Mimics serum adiponectin by forming high molecular weight (HMW) and hexameric species.