Mouse recombinant adiponectin
ENZOALX522059C050
:
- Bal.:50 µG
- Typ proteinu/peptidu:Recombinant
- Druhy:Mouse
- Sekvence:02
- Biologická aktivita:Inhibits serum glucose production in mice. Protects mouse hepatocytes from TNF-α-induced cell death in vitro.
- Pokyny pro rekonstituce:Reconstitute with 100 µl sterile water.
- Proteinová synonyma:ACDC
- Název proteinu/peptidu:Adiponectin
- Čistota:>90% (SDS-PAGE)
- Koncentrace:0,5 mg/ml after reconstitution
- Formulace:Lyophilised. Contains PBS.
Mouse adiponectin (aa 18-247) is fused at the N-terminus to a linker peptide (8 aa) and a FLAG®-tag.
- Produced in HEK 293 cells
Mimics serum adiponectin by forming high molecular weight (HMW) and hexameric species.