Order Entry
Czech Republic
ContactUsLinkComponent
Human recombinant RANKL (soluble)
Human recombinant RANKL (soluble)
Catalog # ENZOALX522012C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  
Human recombinant RANKL (soluble)
Catalog # ENZOALX522012C010
Supplier:  ENZO LIFE SCIENCES
CAS Number:  

Specifications

  • Bal.:
    10 µG
  • Typ proteinu/peptidu:
    Recombinant
  • Druhy:
    Human
  • Sekvence:
    02
  • Biologická aktivita:
    Supports the survival of dendritic cells and osteoclasts.
  • Pokyny pro rekonstituce:
    Reconstitute with 100 µl sterile water. Further dilutions should be made with medium containing 5% fetal calf serum or a carrier protein.
  • Název proteinu/peptidu:
    RANKL (soluble)
  • Čistota:
    >95% (SDS-PAGE)
  • Koncentrace:
    0,1 mg/ml after reconstitution
  • Formulace:
    Lyophilised. Contains PBS.

Specifications

About this item

Produced in HEK 293 cells. The extracellular domain of human RANKL (aa 152-317) is fused at the N-terminus to a linker peptide (6 aa) and a FLAG®-tag.