ContactUsLinkComponent
Human recombinant hepsin
  ENZOALX2017501
 :  
Human recombinant hepsin
  ENZOALX2017501
 :  
  • Bal.:
    1 Vial
  • Typ proteinu/peptidu:
    Recombinant
  • Druhy:
    Human
  • Sekvence:
    02
  • Biologická aktivita:
    Trypsin-like serine protease.
  • Název proteinu/peptidu:
    Hepsin
  • Koncentrace:
    100 nM
  • Formulace:
    Liquid. In 100 mM TRIS, pH 9,0, containing 200 mM sodium chloride and 10% glycerol.
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human hepsin (aa 45-417) is fused at the C-terminus to a V5-His-tag.