Human recombinant hepsin
ENZOALX2017501
:
- Bal.:1 Vial
- Typ proteinu/peptidu:Recombinant
- Druhy:Human
- Sekvence:02
- Biologická aktivita:Trypsin-like serine protease.
- Název proteinu/peptidu:Hepsin
- Koncentrace:100 nM
- Formulace:Liquid. In 100 mM TRIS, pH 9,0, containing 200 mM sodium chloride and 10% glycerol.
Produced in Drosophila Schneider 2 (S2) cells. Recombinant human hepsin (aa 45-417) is fused at the C-terminus to a V5-His-tag.