Cathepsin S drug discovery kit
Catalog # ENZOBMLAK4310001
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Cathepsin S drug discovery kit
- Sekvence:02
- Rozměry:1 kit
Specifications
About this item
A QuantiZyme™ Assay System. The Cathepsin S Drug Discovery Kit is a complete assay system designed to screen inhibitors of cathepsin S. Using a convenient microplate format, cathepsin S activity is monitored fluorimetrically using the substrate provided in the kit.
- Complete system designed to screen modulators of cathepsin S activity
- The assays are performed in a convenient 96-well microplate format
Delivery: Kit contains Cathepsin S enzyme (human) (recombinant), AMC substrate, AMC calibration standard, inhibitor, assay buffer and ½-volume white microplate.