Akt kinase activity kit
Catalog # ENZOADIEKS400A
Supplier: ENZO LIFE SCIENCES
CAS Number:
Specifications
- Bal.:1 KIT
- Popis:Akt kinase activity kit
- Sekvence:02
- Rozměry:96 wells
Specifications
About this item
This is a non-radioactive Akt kinase activity assay, providing a safe, rapid and reliable method for the screening of inhibitors or activators of Akt and for quantitating the activity of Akt in purified or partially purified enzyme preparations. The Akt Kinase activity kit is based on a solid phase enzyme immunoassay (EIA) that utilizes a specific synthetic peptide as a substrate for Akt and a polyclonal antibody that recognizes the phosphorylated form of the substrate. The assay is designed for the analysis of Akt (1, 2, 3) activity in the solution phase.
For the measurement of Akt in partially purified, purified, or crude enzyme preparations from any species.
Ordering information: Contents: Microtiter plate, Antibody, Conjugate, Antibody dilution buffer, Kinase assay dilution buffer, ATP, Active Kinase, Wash buffer concentrate, TMB Substrate, Stop solution 2