ContactUsLinkComponent
LTB4 ELISA kit
  ENZOADI901068
 :  
LTB4 ELISA kit
  ENZOADI901068
 :  
  • Bal.:
    1 KIT
  • Typ testu:
    Competitive
  • Primary antibody reactivity:
    Species independent
  • Cíl:
    LTB₄
  • Popis:
    LTB4 ELISA kit
  • Sekvence:
    02
  • Doba do výsledků:
    4 hrs
  • Citlivost:
    5,63 pg/ml (range 11,7 - 3,000 pg/ml)
For the quantitative determination of LTB4 in culture supernatants, plasma, saliva, and urine from any species. Cited sample types include bronchioalveolar lavage fluid, cell lysate, crevicular fluid, tissue, serum and whole blood.
The LTB4 EIA kit is a colorimetric competitive enzyme immunoassay kit with results in 4 hours. Absorbance is read at 405 nm. Assay up to 39 samples in duplicate with a broad dynamic range that is suitable for a wide variety of sample matrices. Document your results to scientists world-wide with this highly cited kit.
 : Contains GxR Microtiter plate, Conjugate, Antibody, Assay buffer concentrate, Wash buffer concentrate, Standard, pNpp Substrate, Stop solution. Alternative size available: 89141-118 (1x96 wells).