Order Entry
Czech Republic
ContactUsLinkComponent
Anti-Peptide tyrosine tyrosine Mouse Monoclonal Antibody [clone: 4]
Anti-Peptide tyrosine tyrosine Mouse Monoclonal Antibody [clone: 4]
  ENZOBPDABS051041
Anti-Peptide tyrosine tyrosine Mouse Monoclonal Antibody [clone: 4]
  ENZOBPDABS051041

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Peptide tyrosine tyrosine
  • Klonovalita:
    Monoclonal
  • Klon:
    4
  • Spojení:
    Unconjugated
  • ELISA:
    Yes
  • Hostitel:
    Mouse
  • Izotyp:
    IgG2a kappa
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Sekvence:
    03
  • Formulář:
    Liquid
  • Počet aminokyselin:
    3 - 26
  • Skladovací pufr:
    0,01 M phosphate buffer, pH 7,4, containing 0,5 M sodium chloride and 15 mM sodium azide
  • Transportní teplota:
    Blue ice not frozen
  • Imunogen:
    Synthetic peptide corresponding to aa 3-26 of human Peptide YY coupled to carrier
  • Čištění:
    Purified by Protein A/G-affinity chromatography
  • Bal.:
    1 mg

 

 

Type: Primary
Antigen: Peptide tyrosine tyrosine
Clonality: Monoclonal
Clone: 4
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype:
Reactivity: Human