Order Entry
Czech Republic
ContactUsLinkComponent
Anti-TRAIL-R2 Mouse Monoclonal Antibody [clone: DJR2-2]
Anti-TRAIL-R2 Mouse Monoclonal Antibody [clone: DJR2-2]
Katalogové číslo ENZOALX8049130100
Dodavatel:  ENZO LIFE SCIENCES
Anti-TRAIL-R2 Mouse Monoclonal Antibody [clone: DJR2-2]
Katalogové číslo ENZOALX8049130100
Dodavatel:  ENZO LIFE SCIENCES

Specifikace

  • Druh protilátky:
    Primární
  • Název antigenu:
    TNF Receptor superfamily member 10b
  • Symbol antigenu:
    TRAIL-R2
  • Klonovalita:
    Monoclonal
  • Klon:
    DJR2-2
  • Spojení:
    Unconjugated
  • Průtoková cytometrie:
    Yes
  • Hostitel:
    Mouse
  • Imunochemie:
    Yes
  • Izotyp:
    IgG1
  • Sekvence:
    02
  • Formulář:
    Liquid
  • Synonyma antigenu:
    TRAILR2|TRAIL-R2|CD262|Trail receptor 2|TRICK2|KILLER|TRICK2A|TRICKB|ZTNFR9|KILLER/DR5|TRICK2B|DR5
  • Skladovací pufr:
    PBS, pH 7,2. containing 0,09% sodium azide.
  • Skladovací teplota:
    4 °C
  • Transportní teplota:
    Blue Ice Not Frozen
  • Imunogen:
    Recombinant human TRAIL-R2 extracellular domain IgG1 Fc fusion protein.
  • Čištění:
    Purified by affinity chromatography
  • Rozměry protilátky:
    100 µg
  • Bal.:
    100 µG

Specifikace

O tomto produktu

DR5 is a 55kDa member 10B of the TNF receptor superfamily (TNFRSF10B), also known as TRAIL-R2, TRICK2, KILLER, and CD262. It binds the cytotoxic ligand TRAIL and induces apoptosis. The DR5 receptor is broadly expressed on a variety of normal tissues and many tumors. DR5 expression has been reported to be upregulated in human cells by interferon-α, 2-methoxyestradiol, and paclitaxel, and downregulated by adenoviral E3 proteins.

Type: Primary
Antigen: TRAIL-R2
Clonality: Monoclonal
Clone: DJR2-2
Conjugation: Unconjugated
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human