Anti-G Protein Alpha I3 Subunit Rabbit Polyclonal Antibody
ENZOBMLSA1290050
- Druh protilátky:Primární
- Název antigenu:G Protein Alpha I3 Subunit
- Symbol antigenu:G-alpha I3
- Klonovalita:Polyclonal
- Hostitel:Rabbit
- Imunoprecipitace:Yes
- Western blot:Yes
- Sekvence:02
- Formulář:Liquid, in 100 mM potassium phosphate, pH 7,5, containing 140 mM sodium chloride, contains no preservatives
- Čištění:Protein A affinity purified IgG
- Bal.:50 µl
Host: Rabbit
- For Immunoprecipitation, Western Blot
Recognizes human and bovine G protein (alphai3 subunit).
Type: Primary
Antigen: G PROTEIN ALPHA I3 SUBUNIT
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human