Anti-NA+/K+ ATPASE Chicken Polyclonal Antibody
ENZOBMLSA1130200
- Druh protilátky:Primární
- Název antigenu:NA+/K+ ATPASE
- Klonovalita:Polyclonal
- ELISA:Yes
- Hostitel:Chicken
- Western blot:Yes
- Sekvence:02
- Bal.:200 µG
Host: Chicken
- For ELISA, Flow Cytometry, Western Blot
Recognizes mammalian Na+/K+ ATPase.
Type: Primary
Antigen: NA+/K+ ATPASE
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Chicken
Isotype:
Reactivity: Mammals