Zadání objednávky
Czech Republic
ContactUsLinkComponent
Anti-Vasopressin Receptor 2 Rabbit Polyclonal Antibody
Anti-Vasopressin Receptor 2 Rabbit Polyclonal Antibody
  ENZOADI905812100
Anti-Vasopressin Receptor 2 Rabbit Polyclonal Antibody
  ENZOADI905812100

 

  • Druh protilátky:
    Primární
  • Název antigenu:
    Vasopressin Receptor 2
  • Klonovalita:
    Polyclonal
  • ELISA:
    Yes
  • Hostitel:
    Rabbit
  • Imunochemie:
    Yes
  • Reaktivita:
    Human
  • Western blot:
    Yes
  • Sekvence:
    02
  • Formulář:
    Liquid. In PBS containing 50% glycerol and 0,01% sodium azide.
  • Čištění:
    Peptide affinity purified
  • Bal.:
    100 µG

 

 

Arginine vasopressin (AVP) is a nonapeptide hormone produced in magnocellular neurons of the hypothalamus, which functions as an antidiuretic in the kidney and systemically as a regulator of vasoconstriction, blood volume, and blood pressure. AVP acts by binding to a set of specific seven-transmembrane domain-containing G-protein coupled receptors (GPCR) classified into three groups: V1a (V1) receptors regulate vasoconstriction, and are expressed in vascular smooth muscle, kidney, myometrium, bladder, adipocytes, hepatocytes, platelets and testis; V1b (V3) receptors stimulate adrenocorticotropic hormone (ACTH) release in the anterior pituitary, but may also participate in corticotropin-releasing hormone (CRH) release by the hypothalamus; and V2 receptors of the kidney regulate the antidiuretic effects of AVP. The V1b receptor signals through coupling of Gq/11 alpha subunits, activating phospholipase C and release of intracellular Ca2+ via phosphoinositide second messengers. V1b receptor overexpression is a common marker for ACTH-secreting corticotropic adenomas.

Recognizes human, mouse and rat vasopressin receptor V2. Predicted MW of ~41kDa. Higher MW species (~50kDa) also observed by Western blot which reflects post-translational modification of the receptor.

For ELISA, Flow Cytometry, Immunohistochemistry, Western Blot

 : Manufactured by Proteimax.

Type: Primary
Antigen: VASOPRESSIN RECEPTOR 2
Clonality: Polyclonal
Clone:
Conjugation:
Epitope:
Host: Rabbit
Isotype:
Reactivity: Human