Anti-GSK3A Mouse Monoclonal Antibody
Catalog # ENZOADI905759100
Supplier: ENZO LIFE SCIENCES
Specifications
- Druh protilátky:Primární
- Název antigenu:Glycogen Synthase Kinase 3 alpha
- Symbol antigenu:GSK3A
- Klonovalita:Monoclonal
- ELISA:Yes
- Hostitel:Mouse
- Izotyp:IgG1
- Western blot:Yes
- Sekvence:02
- Formulář:Lyophilized from 1 ml of 2× PBS containing 0,09% sodium azide, PEG, and sucrose.
- Synonyma antigenu:GSK-3 alpha|Glycogen synthase kinase-3 alpha|GSK3A|Serine/threonine-protein kinase GSK3A
- Čištění:Thiophilic adsorption and size exclusion chromatography purified
- Bal.:100 µG
Specifications
About this item
Glycogen Synthase Kinase 3 (GSK3) exists in two isoforms, alpha and beta (GSK3α, and GSK3β). GSK3 is an active kinase that becomes phosphorylated by PKB/akt upon growth factor stimulation. Phosphorylation of GSK3α at serine 21 or GSK3β at serine 9, respectively, leads to inhibition of kinase activity of the GSK3.
- For ELISA, Flow Cytometry, Western Blot
Recognizes human, mouse, rat and dog GSK-3alpha.
Reconstitute with 1ml water (15 minutes at room temperature).
Type: Primary
Antigen: GSK-3ALPHA
Clonality: Monoclonal
Clone:
Conjugation:
Epitope:
Host: Mouse
Isotype: IgG1
Reactivity: Human